ContactUsLinkComponent
BulletKit® EGM®-MV Microvascular Endothelial Cell Growth Medium
BulletKit® EGM®-MV Microvascular Endothelial Cell Growth Medium
Catalog # CA71005-534
CAS Number:  
undefined
BulletKit® EGM®-MV Microvascular Endothelial Cell Growth Medium
Catalog # CA71005-534
Supplier Number:  CC-3125
CAS Number:  
Specifications
  • Description:
    EGM®-MV Microvascular endothelial cell growth medium
  • Cat. no.:
    CA71005-534
  • Supplier No.:
    CC-3125
Specifications
About this item
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types.
Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics® Primary Cells.
Media products are supplied in several formats:
Basal Medium – contains no growth factors, cytokines, or supplements
SingleQuots® Kit – growth factors, cytokines, and supplements
BulletKit® – basal medium and SingleQuots® Kit packaged together
The following Endothelial Cell Growth Medium BulletKits® are available:
EGM® BulletKit® – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
EGM®-Plus BulletKit® – Enhanced version of EGM® Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM® Medium.
EGM®-2 BulletKit® – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
EGM®-MV BulletKit® – Bovine Microvascular Cells, contains 5% FBS and BBE. Does not contain exogenous VEGF.
EGM®-2 MV BulletKit® – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
Lonza guarantees the performance of Clonetics®/Poietics® Cells only if appropriate Clonetics®/Poietics® Media and Reagents are used exclusively and the recommended storage and use protocols are followed.