SABM® Small Airway Epithelial Cell Growth Basal Medium
Catalog # CA10065-252
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:SABM® Small airway epithelial cell growth basal medium
- Cat. no.:CA10065-252
- Supplier No.:CC-3119
Specifications
About this item
SABM® is a basal medium sold separately. This is used as a component in the full SAGM® Small Airway Epithelial Cell Growth Medium BulletKit® (catalog number CC-3118) which consists of basal and supplements packaged together. SAGM® BulletKit® is serum-free and has been optimized to grow human small airway epithelial cells(SAEC).
Delivery: Content: 1 × Bottle Basal Medium 500 ml.