BulletKit® SAGM® Small Airway Epithelial Cell Growth Medium
CA10064-352
:
- Description:SAGM® Small airway epithelial cell growth medium
- Environmentally Preferable:
- Cat. no.:CA10064-352
SAGM® BulletKit® is small airway epithelial cell growth medium that is serum-free and has been optimized to grow human small airway epithelial cells (SAEC) from normal and diseased tissue.
Published research shows SAGM® BulletKit® Small Airway Epithelial Cell Growth Medium is also suitable to support co-culture experiments with primary mammary epithelial cells (HMEC) as well as established breast cancer cell lines such as MCF-7 and MDA-MB-231. Refer to the relevant published data in the 'Data' tab.
: Content :
1 × SABM® Basal Medium (CC-3119), 500 ml
1 × SAGM® SingleQuots® Supplement Pack (CC-4124) containing:
1 × Orange Cap Vial with BPE, 2.0 ml
1 × Lilac Cap Vial with Insulin, 0.5 ml
1 × Natural Cap Vial with Hydrocortisone, 0.5 ml
1 × Red Cap Vial with GA-1000, 0.5 ml
1 × Amber Vial with Retinoic Acid, 0.5 ml
1 × Bottle with Fatty Acid Free BSA, 5 ml
1 × Natural Cap Vial with Transferrin, 0.5 ml
1 × Amber Vial with Triiodothyronine, 0.5 ml
1 × Amber Vial with Epinephrine, 0.5 ml
1 × Green Cap Vial with hEGF, 0.5 ml
1 × SABM® Basal Medium (CC-3119), 500 ml
1 × SAGM® SingleQuots® Supplement Pack (CC-4124) containing:
1 × Orange Cap Vial with BPE, 2.0 ml
1 × Lilac Cap Vial with Insulin, 0.5 ml
1 × Natural Cap Vial with Hydrocortisone, 0.5 ml
1 × Red Cap Vial with GA-1000, 0.5 ml
1 × Amber Vial with Retinoic Acid, 0.5 ml
1 × Bottle with Fatty Acid Free BSA, 5 ml
1 × Natural Cap Vial with Transferrin, 0.5 ml
1 × Amber Vial with Triiodothyronine, 0.5 ml
1 × Amber Vial with Epinephrine, 0.5 ml
1 × Green Cap Vial with hEGF, 0.5 ml