SingleQuots® HCM® Kit
Catalog #: CA10063-992
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:HCM® kit
- Cat. no.:CA10063-992
Specifications
About this item
The HCM® SingleQuots® Kit consists of individual vials containing the key labile ingredients required to combine with the HBM® Basal Medium(Sold Seperately, CC-3199)in support of primary hepatocyte cultures. Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.
- Optimized for Hepatocyte cell health
- Compatible with all species Hepatocytes
Delivery: Content: 1 × HCM® Singlequots® Growth Supplement Pack (CC-4182).
Customers Who Bought This Also Bought