Rat Brain Striatum Astrocytes
CA10064-114
:
- Description:Rat brain striatum astrocytes
- Size:≥1 million cells
- Cat. no.:CA10064-114
Ready-to-use Clonetics® Rat Brain Striatum Astrocytes are obtained from rat brain, passaged once and prepared as cell suspensions for shipment on dry ice.
- Ready-To-Use
- No tissue dissection or animal handling required
- Increased data reproducibility
- Characteristic morphology and staining tests confirm tissue origin and cell type
- Negative for mycoplasma, bacteria, yeast and fungi
Each vial contains approximately 1 million cells. Astrocytes can be easily thawed and cultured. Astrocytes can be harvested and re-plated or stored frozen for later use. The cells are shipped overnight on dry ice.
Recommended medium: AGM® BulletKit® (CC-3186)
Applications: Neurogenesis, Neurotransmitter function, Gene expression, Signaling pathways, Electrophysiology, Neurotoxicity, Drug/Compound screening, Advanced cell culture models.
: Content: ≥1000000 cryopreserved cells per ampule. Certificate of Analysis (CoA) is available for every batch.