Orders LinkContactUsLinkComponent
hMSC - Human Mesenchymal Stem Cells
hMSC - Human Mesenchymal Stem Cells
hMSC - Human mesenchymal stem cells, 750000 cells
Catalog # CA10064-080
Supplier #:  PT-2501
hMSC - Human Mesenchymal Stem Cells
Catalog # CA10064-080
CAS Number:  
hMSC - Human Mesenchymal Stem Cells
hMSC - Human Mesenchymal Stem Cells
hMSC - Human mesenchymal stem cells, 750000 cells
CAS Number:  
Price
$2,273.58
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Poietics® Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) are isolated from normal (non-diabetic) adult human bone marrow withdrawn from bilateral punctures of the posterior iliac crests of normal volunteers.
  • All cells test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots
Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) have been reported to differentiate down many different lineages including chondrogenic, osteogenic, adipogenic and neural. Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) have been cryopreserved after the second passage and are suggested to be used by no later than the fifth passage. Cryopreserved Poietics® Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) are guaranteed to have at least 0.75 million viable cells per vial, differentiate down the adipogenic, chondrogenic, and osteogenic liniages when cultured in the recommended differentiation medium, to express CD29, CD44, CD105, and CD166 and to not express CD14, CD34, and CD45. All lots of this product manufactured after January 1, 2015 are also guaranteed to express CD90 and CD73 and to not express HLA-DR or CD19.
Lonza guarantees the performance of Clonetics®/Poietics® cells only if appropriate Clonetics®/Poietics® media and reagents are used exclusively and the recommended storage and use protocols are followed.
Delivery information: A Certificate of Analysis is provided for each cell lot purchased.
Specifications
Specifications
  • Description:
    hMSC - Human mesenchymal stem cells
  • Size:
    750000 cells