Order Entry
Canada
ContactUsLinkComponent
Mouse C57 Brain Cortex Neurons
Mouse C57 Brain Cortex Neurons
Catalog # CA10064-064
CAS Number:  
undefined
Mouse C57 Brain Cortex Neurons
Catalog # CA10064-064
Supplier Number:  M-CX-300
CAS Number:  

Specifications

  • Description:
    Mouse C57 brain cortex neurons
  • Size:
    ≥4 million cells
  • Cat. no.:
    CA10064-064
  • Supplier No.:
    M-CX-300

Specifications

About this item

Ready-to-use Clonetics® Mouse Brain C57 Cortex Neurons (M-CX-300) are isolated from Mouse C57 black brain (E14, 15). These are sold as cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods, and are ready for immediate culture.

  • Ready-To-Use
  • No tissue dissection or animal handling required
  • Increased data reproducibility
  • Characteristic morphology and staining tests confirm tissue origin and cell type
  • Negative for mycoplasma, bacteria, yeast and fungi

Each vial of cortex cells contains approximately 4 million viable cells. This will seed into 10 or more wells of a 24-well plate and 50 or more wells of a 96-well plate using the recommended plating densities and medium. After approximately 4 days in culture, the cells will form a neurite network and by the 7th day, debris will be minimal.

Recommended medium: PNGM® BulletKit® (CC-4461).

Applications: Neurogenesis, Neurotransmitter function, Gene expression, Signaling pathways, Electrophysiology, Neurotoxicity, Drug/Compound screening, Advanced cell culture models.

Delivery: Content: ≥4000000 cryopreserved cells per ampule. Certificate of Analysis (CoA) is available for every batch.