HMVEC-C – Human Cardiac Microvascular Endothelial Cells
Catalog # CA10064-376
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:HMVEC-C – human cardiac microvascular endothelial cells
- Size:≥500000 cells
- Cat. no.:CA10064-376
Specifications
About this item
Human Cardiac Microvascular Endothelial Cells (HMVEC-C) are isolated from normal and diabetic donors.
- Guaranteed through 15 population doublings
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Clonetics® Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system. Cryopreserved Human Cardiac Microvascular Endothelial Cells are shipped in third passage.
Cryopreserved Human Cardiac Microvascular Endothelial Cells (CC-7030) are guaranteed through 10 population doublings, express CD31/105, von Williebrand Factor VIII, and are positive for acetyated low density lipoprotein uptake.
Delivery: Content: 1× Cryopreserved ampule of HMVEC-C containing ≥500000 cells. A Certificate ofAnalysis is provided for each cell lot purchased.