Orders LinkContactUsLinkComponent
HMVEC-Bd – Human Bladder Microvascular Endothelial Cells
HMVEC-Bd – Human Bladder Microvascular Endothelial Cells
HMVEC-Bd – Human bladder microvascular endothelial cells, ≥500000 cells
Catalog # CA10064-374
Supplier #:  CC-7016
HMVEC-Bd – Human Bladder Microvascular Endothelial Cells
Catalog # CA10064-374
CAS Number:  
HMVEC-Bd – Human Bladder Microvascular Endothelial Cells
HMVEC-Bd – Human Bladder Microvascular Endothelial Cells
HMVEC-Bd – Human bladder microvascular endothelial cells, ≥500000 cells
CAS Number:  
Price
$2,289.03
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Human Bladder Microvascular Endothelial Cells include cells isolated from the microvasculature of normal tissue surrounding the bladder. Clonetics® Endothelial Cells and Media are convenient and easy to use, allowing the researcher to focus on results.
  • All cells test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots
Clonetics® Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system. Cryopreserved cells are shipped in third passage.
Cryopreserved Human Bladder Microvascular Endothelial Cells are guaranteed through 10 population doublings, express CD31/105, von Williebrand Factor VIII, and are positive for acetyated low density lipoprotein uptake.
Delivery information: Content: 1 × Cryopreserved Ampule with ≥500000 cells/vial. A Certificate of Analysis is provided for each cell lot purchased.
Specifications
Specifications
  • Description:
    HMVEC-Bd – Human bladder microvascular endothelial cells
  • Size:
    ≥500000 cells
Specifications