Specifications
- Description:NHDC - Human dendritic cells
- Size:2.5 million cells
- Cat. no.:CA10064-282
Specifications
About this item
Dendritic Cells (DCs) act as the messenger cells of the immune system by processing antigens from pathogens and presenting the antigens to T cells to initiate an immune response.
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Poietics® Normal Human Dendritic Cells are tested for a panel of specific surface markers, including CD11c+, CD86+, CD80+. HLADR+, and CD14- to provide detailed characterization of the cells. These cells can survive up to 7 days in culture with the proper cytokines (IL-4 and GM-CSF) to maintain the cells as immature dendritic cells (iDCs). Cells can be further differentiated to mature dendritic cells with the addition of TNF-α and IL-1β.
Applications: Antigen presentation and uptake Primary Research Pharmacology Drug-induced immunosuppression of cytokine secretion.
Cryopreserved Poietics® Normal Human Dendritic Cells (NHDC) are guaranteed to contain ≥3 million viable cells per ampoule. Expression levels are tested for CD80, CD86, HLA-DR, CD11c, and CD14 and provided for information only. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased. Recommended Medium: LGM-3® Lymphocyte Growth Medium + 50 ng/ml GM-CSF + 50 ng/ml IL-4.