CASMC – Human Coronary Artery Smooth Muscle Cells
CA10064-272
:
- Description:CASMC – Human coronary artery smooth muscle cells
- Size:≥500000 cells
- Cat. no.:CA10064-272
- Supplier No.:CC-2583
Human Vascular Smooth Muscle Cells come from multiple tissue types including Aorta (AoSMC), Coronary Artery (CASMC), Pulmonary Artery (PASMC), and Umbilical Artery (UASMC).
- Applications: Stroke, Diabetes, Vascular research, Artherosclerosis,Hypertension
Clonetics® Human Aortic Smooth Muscle Cells are guaranteed for 15 population doublings and to stain positive for alpha smooth muscle actin and negative for von Willebrand Factor VIII after differentiation. All cells test negative for mycoplasma, bacteria, yeast and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
: Content: 1 × Cryopreserved Ampule with ≥500000 cells/vial. Cryopreserved cells are shipped in 3rd passage. A Certificate of Analysis (CoA) is provided for each cell lot purchased.