Order Entry
Canada
ContactUsLinkComponent
PASMC - Human Pulmonary Artery Smooth Muscle Cells
PASMC - Human Pulmonary Artery Smooth Muscle Cells
Catalog # CA10064-270
CAS Number:  
undefined
PASMC - Human Pulmonary Artery Smooth Muscle Cells
Catalog # CA10064-270
Supplier Number:  CC-2581
CAS Number:  

Specifications

  • Description:
    PASMC - Human pulmonary artery smooth muscle cells
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-270
  • Supplier No.:
    CC-2581

Specifications

About this item

Human Pulmonary Artery Smooth Muscle Cells (PASMC) are shipped in 3rd passage. Guaranteed for 15 population doublings and to stain positive for alpha smooth muscle actin and negative for von Willebrand Factor VIII after differentiation.

  • All cells test negative for mycoplasma, bacteria, yeast and fungi
  • HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots

Applications: Stroke, Diabetes, Vascular research, Artherosclerosis, Hypertension.

Delivery: Content: 1 × Cryopreserved ampule PASMC containing ≥500000 cells/vial. A Certificate of Analysis (CoA) is provided for each cell lot purchased.