BSMC – Human Bronchial Smooth Muscle Cells
CA10064-264
:
- Description:BSMC – Human bronchial smooth muscle cells
- Size:≥500000 cells
- Cat. no.:CA10064-264
- Supplier No.:CC-2576
Human Bronchial Smooth Muscle Cells (BMSC) are isolated from the major bronchia of normal tissue.
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Cryopreserved Clonetics® Human Bronchial Smooth Muscle cells are guaranteed through 15 population doublings and to stain positive foralpha smooth muscle actin and negative for von Willebrand Factor VIII.
: Content: 1 × Cryopreserved ampule BSMC containing ≥500000 cells/vial. Cells are shipped in passage 2. A Certificate of Analysis is provided for each cell lot purchased.