PrEC – Human Prostate Epithelial Cells
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Prostate cells provide a glandular function in the body by generating fluid which serves several functions in reproduction. Lonza's Human Prostate Cells are available from epithelial, smooth muscle, and stromal tissue. Each cell type has an optimized growth media for best results in culture.
- Guaranteed through 15 population doublings
- Test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Prostate Epithelial Cells (PrEC) are guaranteed through 15 population doublings and stain positive for cytokeratin (clone 8.13). Cryopreserved PrEC are shipped in first or second passage.
Applications: Benign prostatic hyperplasia (BPH), Prostitis, Prostate cancer, Procreation research, Drug discovery and Male prostate physiology.
Delivery information: Content: 1 × Cryopreserved ampule PrEC containing ≥500000 cells. A Certificate of Analysis (CoA) is provided for each cell lot purchased.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:PrEC – Human prostate epithelial cells
- Size:≥500000 cells