SAEC – Human Small Airway Epithelial Cells
CA10064-220
:
- Description:SAEC – Human small airway epithelial cells
- Size:≥500000 cells
- Cat. no.:CA10064-220
- Supplier No.:CC-2547
Lonza's Human Small Airway Epithelial Cells (SAEC) are isolated from normal human lung tissue the distal portion of the lung in the 1mm bronchiole area.
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Cryopreserved SAECs or Alveolar Cells are guaranteed through 15 population doublings and stain positive for cytokeratin 19.SAECs or Alveolar Cells labeled CC-2547S are suitable for air-liquid interface studies and are guaranteed for differentiation marked by cilia formation, mucin production, and TEER levels. Cells from various diseased donors are also available.
: Content: 1 × Cryopreserved ampule SAEC containing ≥500000 cells. Certificate of Analysis provided for every cell lot.