To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:S-100/S100A1
- Clonality:Polyclonal
- Gene ID:6271
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Chicken,Human,Bovine,Dog,Rat,Mouse
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Storage temperature:Store at –20 °C for 12 months.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Specifications
About this item
S-100 is group of solubilities, the relatively low molecular mass, the high acidic protein, is the neuroglia cell specificity protein, may also exist in the nerve fiber. In the research intestines nervous system's shape characteristic and the structural feature, S-100 has the good expressive function. The S-100 protein its physiological action was not still clear, possible and in the cell the calcium ion medium's control action concerns. The S-100 protein may exist in the neurinoma, becomes the cartilage cytoma, the chondrosarcoma, the osteosarcoma, the neuroglia lump, Lang Gehan the Si cytoma, some cancers and the melanocyte cell tumor and so on. In the immunity group, damages except the malignant melanoma skin, the S-100 protein also increases in the benign melanocyte cell mole, for example: The blue nevus, the ordinary day after tomorrow mole, the freckle type mole, the Spitz mole, the gas globuli cell mole, the pigment spindle-shaped cell mole and in the maldevelopment mole will express. Also has reported that the S-100 protein's dyeing intensity and tumour cell's melanocyte content assumes the inverse correlation. The knot contracts the organization proliferative or kisses the nerve the melanoma undyeable color. SUBUNIT: Dimer of either two alpha chains, or two beta chains, or one alpha and one beta chain.
Type: Primary
Antigen: S-100/S100A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken