ContactUsLinkComponent
D-SAEC-COPD – Diseased Small Airway Epithelial Cells – COPD
D-SAEC-COPD – Diseased Small Airway Epithelial Cells – COPD
Catalog # CA10064-340
CAS Number:  
undefined
D-SAEC-COPD – Diseased Small Airway Epithelial Cells – COPD
Catalog # CA10064-340
Supplier Number:  CC-2934
CAS Number:  
Specifications
  • Description:
    D-SAEC-COPD – Diseased small airway epithelial cells – COPD
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-340
Specifications
About this item
Cystic fibrosis is a disease that causes thick, sticky mucus to build up in the lungs, digestive tract, and other areas of the body. It is one of the most common chronic lung diseases in children and young adults.
The disease is the most common, deadly, inherited disorder affecting Caucasians in the United States. The disease eventually worsens to the point where the person is disabled. Today, the average life span for people with CF who live to adulthood is approximately 37 years, a dramatic increase over the last three decades.
Human Small Airway Epithelial Cells (SAEC) are isolated from cystic fibrosis human lung tissue. Cryopreserved Clonetics® SAECs are guaranteed through 15 population doublings and stain positive for cytokeratin 19. All cells testnegative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B andhepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved ampule with ≥500000 cells/vial and Certificate of Analysis is provided for each cell lot purchased.