SAEC - Small Airway Epithelial Cells for S-ALI® Small Airway Air-Liquid-Interface Medium
Catalog # CA10064-222
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:SAEC - Small Airway Epithelial Cells for S-ALI® small airway air-liquid-interface medium
- Size:≥500000 cells
- Cat. no.:CA10064-222
Specifications
About this item
Human Small Airway Epithelial Cells (SAEC) are isolated from normal human lung tissue the distal portion of the lung in the 1mm bronchiole area. Contact our Scientific Support team for additional details.
Cryopreserved Clonetics® SAECs are guaranteed through 15 population doublings and stain positive for cytokeratin 19.SAECs labeled CC-2547S are suitable for air-liquid interface studies and are guaranteed for differentiation marked by cilia formation, mucin production, andTEER levels. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B andhepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved ampule with ≥500000 cells/vial and Certificate of Analysis is provided for each cell lot purchased.