Nucleofector® SG Cell Line 96-well Kit
Catalog # CA10064-130
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:SG cell line 96-well Nucleofector® kit
- Cat. no.:CA10064-130
- No. of Reactions:96
- Supplier No.:V4SC-3096
Specifications
About this item
The SG cell line 96-well Nucleofector® kit is one of our three kits suited for medium-throughput transfection of cell lines when working with the 4D-Nucleofector® 96-well unit.
- High transfection efficiencies in difficult-to-transfect cell lines
- Compatible with liquid handling systems for automated high-throughput electroporation
- Conditions are transferable between 4D-Nucleofector® System, 4D-Nucleofector® 96-well Unit and 384-well Nucleofector® System
This kit has been determined to be optimal for transfecting following cell types:
Ba/F3
Colo-205
H4
NIH/3T3
Raji, Ramos
THP-1
WEHI
The kit is available in different variants for either 1×96 or 10×96 reactions.
Applications:
Transfection of low cell numbers in the range of 5×10⁴ to 1×10⁶ cells.
High-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR libraries.
Delivery: Content:
2.25 ml SG Cell Line Nucleofector® solution.
0.5 ml Supplement 1.
50 µg pmaxGFP® vector (1 μg/μl in 10 mM Tris pH 8.0).
1×96-well Nucleocuvette® Plate (20 μl).
2.25 ml SG Cell Line Nucleofector® solution.
0.5 ml Supplement 1.
50 µg pmaxGFP® vector (1 μg/μl in 10 mM Tris pH 8.0).
1×96-well Nucleocuvette® Plate (20 μl).