Order Entry
Canada
ContactUsLinkComponent
Nucleofector® SF Cell Line 96-well Kit
Nucleofector® SF Cell Line 96-well Kit
Catalog # CA10064-128
CAS Number:  
undefined
Nucleofector® SF Cell Line 96-well Kit
Catalog # CA10064-128
Supplier Number:  V4SC-2096
CAS Number:  

Specifications

  • Description:
    SF cell line 96-well Nucleofector® kit
  • Cat. no.:
    CA10064-128
  • No. of Reactions:
    96
  • Supplier No.:
    V4SC-2096

Specifications

About this item

The SF Cell Line 96-well Nucleofector® kit is one of our three kits suited for medium-throughput transfection of cell lines when working with the 4D-Nucleofector® 96-well unit.

  • High transfection efficiencies in difficult-to-transfect cell lines
  • Compatible with liquid handling systems for automated high-throughput electroporation
  • Conditions are transferable between 4D-Nucleofector® System, 4D-Nucleofector® 96-well Unit and 384-well Nucleofector® System

This kit has been determined to be optimal for transfecting following cell types:
293-F (FreeStyle), 32D
A-20, A2058, A375, A431, A549, AGS, ARPE-19
BetaTC/6, BxPC-3
C6, CCRF-CEM, CHO-K1, CHO-S (Invitrogen), Cos-1
EL4
FDC-P1
H9c2 (2-1), HEK293, Hel9217, HepG2, High 5, HL-60, HMC-1-8, HT-1080, HT-29, Hut-78, HUV-EC-C
IMR-32
J774A.1
K-562
KG-1
L-428, LNCaP
MDA-MB-453, MDBK, Molt-4, MV-4-11
Nalm-6, NB-4, NCI-H1299 [H1299], Neuro-2a [N2a], NG108-15, NS0
P19, P815, PC-12, PC-3
Raw 264.7, RBL-1, RBL-2H3, Reh, Rin m5F, RPMI 8226
S2, S49, SaOS, SF9, SH-SY5Y, SK-BR-3, SK-N-AS, SK-N-MC, SK-N-SH, SK-OV-3, Sp2/0-Ag14, SW480
T2, T84, TF-1
U266B1, U-937
Vero
The respective cell type-specific Optimized Protocols or recommendations are available under ""Instructions"" or in our knowledge database. In these Optimized Protocols the best Nucleofection conditions are indicated. In addition, we share our experience and knowledge for treatment of individual cell types.

The kit is available in different variants for either 1 x 96 or 10 x 96 reactions.

Applications:
Transfection of low cell numbers in the range of 5×10⁴ to 1×10⁶ cells.
High-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR libraries.

Delivery: Content:
2.25 ml SF cell line Nucleofector® solution.
0.5 ml Supplement 1.
50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0).
1×96-well Nucleocuvette® Plate (20 μl).