Anti-IFN gamma Rabbit Polyclonal Antibody
Catalog # 77437-058
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IFN gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human
- Size:100 µl
- Gene ID:3458
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:17 kDa
- Storage temperature:Store at –20 °C for 12 months.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. no.:77437-058
- Packaging:Vial
Specifications
About this item
Mammalian Interferon gamma is mainly produced by T lymphocytes and NK cells. It is a pleiotropic cytokine involved in the regulation of nearly all phases of immune and inflammatory responses,including the activation, growth and differentiation of T cell, B cells, macrophages, NK cells and other cell types such as endothelial cells and fibroblasts. It has weak antiviral and antiproliferative activity, and poteniates the antiviral and anti tumor effects of IFN alpha/beta (type I interferon). It is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. Labile at pH 2.
Type: Primary
Antigen: IFN gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human