FGM®-3 Cardiac Fibroblast Growth Medium-3 BulletKit®
Catalog # CA10064-038
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:FGM®-3 cardiac fibroblast growth medium-3 BulletKit®
- Cat. no.:CA10064-038
Specifications
About this item
FGM®-3 fibroblast growth medium-3 was developed specifically for culturing primary cardiac fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM®-3 medium contains contains 10% serum.
Clonetics® NHCF-V and -A are guaranteed through 5 population doublings and express ≥90% collagen I and ≤10% Von Willibrand factor VIII after thaw. Clonetics® AoAF are guaranteed through 10 population doublings and are negative for alpha smooth muscle actin. All cells test negative for mycoplasma, bacteria, yeast and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery: Content:
1×FBM® basal medium (CC-3131), 500 ml.
1×FGM®-3 SingleQuots® supplement pack (CC-4525).
1×Lilac cap vial with insulin, 0.50 ml.
1×Gray cap vial with hFGF-B, 0.50 ml.
1×Red cap vial with GA-1000, 0.50 ml.
1×Bottle FBS, 50.00 ml.
A Certificate of Analysis (CoA) is provided for each cell lot purchased.
1×FBM® basal medium (CC-3131), 500 ml.
1×FGM®-3 SingleQuots® supplement pack (CC-4525).
1×Lilac cap vial with insulin, 0.50 ml.
1×Gray cap vial with hFGF-B, 0.50 ml.
1×Red cap vial with GA-1000, 0.50 ml.
1×Bottle FBS, 50.00 ml.
A Certificate of Analysis (CoA) is provided for each cell lot purchased.