Order Entry
Canada
Orders LinkContactUsLinkComponent
Nucleofector® P1 Primary Cell 4D-X Kit S

Nucleofector® P1 Primary Cell 4D-X Kit S

P1 Primary Cell 4D-Nucleofector® X kit S, 32
Catalog # CA10064-234
Supplier #:  V4XP-1032
Nucleofector® P1 Primary Cell 4D-X Kit S
Catalog # CA10064-234
CAS Number:  
Nucleofector® P1 Primary Cell 4D-X Kit S
Nucleofector® P1 Primary Cell 4D-X Kit S

P1 Primary Cell 4D-Nucleofector® X kit S, 32

CAS Number:  
Price
$901.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The P1 primary cell 4D-Nucleofector® X kit S is one of our five 4D-Nucleofector® kits suited for transfecting primary cells in the 20 µl 16-well Nucleocuvette® Strip format when working with the 4D-Nucleofector® X unit.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
  • Conditions are transferable between 4D-Nucleofector® System, 96-well Shuttle® System and 384-well Nucleofector® System

P1 kit has been determined to be optimal for transfecting following cell types:
ADSC undiff. (human)
MSC (human)
Preadipocytes (human)
PrEC (human)

Applications:
Transfection of low cell numbers in the range of 5×10⁴ to 1×10⁶ cells.
Medium throughput applications.
Optimization of transfection conditions e.g. substrate titrations.

Delivery information: Content:
1×0.675 ml P1 primary cell Nucleofector® solution.
1×0.15 mL Supplement 1.
50 µg pmaxGFP® vector (1 μg/μl in 10 mM Tris pH 8.0).
2×16-well Nucleocuvette® strips (20 μl).

Specifications

Specifications

  • Description:
    P1 Primary Cell 4D-Nucleofector® X kit S
  • No. of reactions:
    32

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.