Order Entry
Canada
ContactUsLinkComponent
D-HSMM – Diseased Human Skeletal Muscle Myoblasts – Diabetes Type I
D-HSMM – Diseased Human Skeletal Muscle Myoblasts – Diabetes Type I
Catalog # CA10064-296
CAS Number:  
undefined
D-HSMM – Diseased Human Skeletal Muscle Myoblasts – Diabetes Type I
Catalog # CA10064-296
Supplier Number:  CC-2900
CAS Number:  

Specifications

  • Description:
    D-HSMM – Diseased Human Skeletal Muscle Myoblasts – diabetes type I
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-296

Specifications

About this item

Clonetics® Diabetic type I and type II Human Skeletal Muscle Myoblasts (HSMM) are isolated from donors diagnosed with either diabetes type I or diabetes type II.

Diseased cells are tested in the same SkGM®-2 skeletal muscle growth media as the normal cells allowing for easy comparisons between diseased and normal tissue.

Clonetics® Diabetic donor HSMMs are tested are guaranteed to differentiate to form multinucleated myotubes in culture which stain positive for desmin. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.

Applications: Cellular development, differentiation, insulin uptake or resistance, tissue repair, metabolic effects of diabetes, myotube development.

Delivery: Content: 1×cryopreserved ampule with ≥500000 cells/vial.