RtEGM® Retinal Pigment Epithelial Cell Growth Medium BulletKit®
CA10063-842
:
- Description:RtEGM® retinal pigment epithelial cell growth medium Bulletkit®
- Cat. no.:CA10063-842
- Supplier No.:00195409
RtEGM® Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum.
Applications: Age-related macular degeneration, retinitis pigmentosa, diabetic retinopathy, photoreceptor degeneration, gene expression, cellular interactions.
: Content:
1×RtEBM® Basal medium (00195406), 200 ml.
1×RtEGM® SingleQuots® Supplement pack (00195407)
1×Bottle L-glutamine, 4.00 ml.
1×Gray cap vial with hFGF-B, 1.00 ml
1×Bottle FBS, 4.00 ml.
1×Red cap vial with GA-1000, 0.20 ml.
1×RtEBM® Basal medium (00195406), 200 ml.
1×RtEGM® SingleQuots® Supplement pack (00195407)
1×Bottle L-glutamine, 4.00 ml.
1×Gray cap vial with hFGF-B, 1.00 ml
1×Bottle FBS, 4.00 ml.
1×Red cap vial with GA-1000, 0.20 ml.