RtEGM® Retinal Epithelial Cell Growth Medium SingleQuots® Supplements and Growth Factors
CA10063-840
:
- Description:RtEGM® Retinal epithelial cell growth medium SingleQuots® supplements and growth factors
- Cat. no.:CA10063-840
RtGM® SingleQuot® kit are components sold separately and does not include the basal medium. These are part of the RtEGM® Retinal Epithelial Cell Growth Medium Bulletkit® (Catalog Number 195409).
The full RtEGM® BulletKit® consisting of basal medium and supplements also available.
RtEGM® Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum.
Applications: Age-related macular degeneration, retinitis pigmentosa, diabetic retinopathy, photoreceptor degeneration, gene expression, cellular interactions.
: Content: 1×bottle with L-glutamine 4.00 ml, 1×gray cap vial with hFGF-B 1.00 ml, 1×bottle FBS 4.00 ml, 1×red cap vial with GA-1000 0.20 ml.