- Description:D-HBE-COPD – Diseased Human Bronchial/Tracheal Epithelial cells – COPD
- Size:≥500000 cells
- Cat. no.:CA10063-834
COPD airway cells include lung fibroblasts, bronchial epithelial and bronchial smooth muscle cells. Bronchial epithelial cells from COPD donors are isolated from epithelial lining of airways above bifurcation of the lungs.
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Cryopreserved diseased human bronchial epithelial cells from COPD donors test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Our catalog number 00195275S are suitable for air-liquid interface studies and are guaranteed to promote full differentiation of the airway epithelium expressed by the formation of a polarized epithelium with good barrier function (transepithelial resistance), secretory phenotype (mucin secretion) and ciliogenesis. Differentiation usually will occur by day 22 in culture.
Applications: Gene expression pathways, phenotypic changes to compounds, transfect diseased cells, co-culture with immune cells, airway remodeling studies, cellular interactions.