Specifications
- Description:B-ALI® Bronchial Air-Liquid interface medium Bulletkit®
- Cat. no.:CA10063-818
Specifications
About this item
B-ALI® Bronchial air liquid interface medium Bulletkit® provides scientists with a unique tool to investigate new areas of airway epithelial research in a more physiologically relevant way.
By using an air-liquid interface culture, the next generation tool of in vitro respiratory research, scientists can explore bronchial airway epithelial cell differentiation and apply therapies in novel ways.
The B-ALI® Bullekit® is made up of an air liquid interface Growth and differentiation medium SingleQuots® Kit, Growth basal medium and differentiation basal medium.
Our B-ALI® BulletKit® promotes full differentiation of the airway epithelium expressed by the formation of a polarized epithelium with good barrier function (transepithelial resistance), secretory phenotype (mucin secretion) and ciliogenesis. When used with our guaranteed bronchial epithelial cells(CC-2540S).
1×B-ALI® basal media (00193516), 250 ml.
1×B-ALI® differentiation media (00193517), 500 ml.
1×B-ALI® SingleQuots® supplement pack (00193515).
1×Natural cap vial with transferrin, 0.75 ml.
1×Amber vial with B-ALI-Inducer, 1.00 ml.
1×Bottle BPE, 3.00 ml.
1×Amber vial with epinephrine, 0.75 ml.
1×Red cap vial with GA-1000, 0.75 ml.
1×Green cap vial with hEGF, 0.75 ml.
1×Natural cap vial with hydrocortisone, 0.75 ml.
1×Lilac cap vial with insulin, 0.75 ml.
1×Amber vial with retinoic acid, 0.75 ml.
1×Amber vial with triiodothyronine, 0.75 ml.