Rat Neonatal Retinal Cells
Catalog # CA10064-174
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Rat neonatal retinal cells
- Size:≥200000 cells
- Cat. no.:CA10064-174
Specifications
About this item
Clonetics® Neonatal rat retinal cells [P3-4] are ready-to-use, high quality primary cells from Sprague Dawley rats. Cells are prepared by standardized methods and are ready for immediate culture upon thaw. Cells are cryopreserved with ≥0.2 M viable cells per ampule.
- Ready-to-use
- No tissue dissection or animal handling required
- Increased data reproducibility
- Characteristic morphology and staining tests confirm tissue origin and cell type
- Negative for mycoplasma, bacteria, yeast and fungi
Cells are characterized by immunostaining for neuron specific class III β-tubulin (Tuj-1), specific neuronal protein gene product (PGP 9.5), ganglion cell marker, Thy1.1 and GFAP.
Recommended medium: PNGM® BulletKit®
Applications: General ophthalmic research, posterior segment disease, inflammation, degeneration, neoplasms, drug delivery, cell therapies, toxicology and cytotoxicity, gene expression.
Delivery: Content: ≥200000 cryopreserved cells per ampule.