KGM® Gold Keratinocyte Growth Medium SingleQuots® Supplements and Growth Factors
Catalog # CA10063-798
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:KGM® gold keratinocyte growth medium SingleQuots® supplements and growth factors
- Cat. no.:CA10063-798
Specifications
About this item
KGM® Gold SingleQuot® kit are components sold separately. These are part of the KGM® Gold Keratinocyte Growth Medium Bulletkit® (Catalog Number 192060). The full KGM® Gold Bulletkit® consisting of basal medium and supplements can be purchased together using the catalog number 192060.
KGM® Gold BulleKit® is a new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
Delivery: Content:
1×Natural cap vial with hydrocortisone 0.50 ml
1×Natural cap vial with transferrin 0.50 m
1×Amber vial with epinephrine 0.25 ml
1×red cap vial with GA-1000 0.50 ml
1×orange cap vial with BPE 2.00 ml
1× green cap vial with hEGF 0.50 ml
1× lilac cap vial with Insulin 0.50 ml
1×Natural cap vial with hydrocortisone 0.50 ml
1×Natural cap vial with transferrin 0.50 m
1×Amber vial with epinephrine 0.25 ml
1×red cap vial with GA-1000 0.50 ml
1×orange cap vial with BPE 2.00 ml
1× green cap vial with hEGF 0.50 ml
1× lilac cap vial with Insulin 0.50 ml