Order Entry
Canada
ContactUsLinkComponent
Nucleofector® Basic Kit for Primary Mammalian Fibroblasts
Nucleofector® Basic Kit for Primary Mammalian Fibroblasts
Catalog # CA10064-638
CAS Number:  
undefined
Nucleofector® Basic Kit for Primary Mammalian Fibroblasts
Catalog # CA10064-638
Supplier Number:  VPI-1002
CAS Number:  

Specifications

  • Description:
    Basic Nucleofector® kit for primary mammalian fibroblasts
  • Cat. no.:
    CA10064-638
  • No. of Reactions:
    25
  • Supplier No.:
    VPI-1002

Specifications

About this item

In addition to various cell type-specific kits for primary fibroblasts, we also provide a cell group-specific basic Nucleofector® kit.

  • Optimization can be performed within 1 experiment
  • Detailed protocols guiding through the optimization procedure
  • Optional result fine tuning with help of our Scientific Support Team
  • Transfection efficiencies up to 90%
  • Viabilities up to 98%

This kit is suited to determine the optimal transfection conditions for any mammalian fibroblast type that lacks a cell type specific Optimized Protocol for the Nucleofector® 2b device (or older generations I or II). In a first experiment a selection of different Nucleofector® programs is tested in combination with the basic Nucleofector® solution for fibroblasts.

Applications: kit suited for fibroblasts from different mammalian species and various organs, already tested for macaque dermal fibroblasts bovine fibroblasts human colon myofibroblasts mouse lung fibroblasts etc.

Delivery: Content: 2.25 ml Nucleofector® solution for primary mammalian fibroblasts, 0.5 ml supplement 1, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mM Tris pH 8.0), 25 aluminum cuvette (100 µL), 25 single use pipettes.