Order Entry
Canada
Orders LinkContactUsLinkComponent
Nucleofector® Basic Kit for Primary Mammalian Endothelial Cells

Nucleofector® Basic Kit for Primary Mammalian Endothelial Cells

Basic Nucleofector® kit for primary mammalian endothelial cells, 25
Catalog # CA10064-636
Supplier #:  VPI-1001
Nucleofector® Basic Kit for Primary Mammalian Endothelial Cells
Catalog # CA10064-636
CAS Number:  
Nucleofector® Basic Kit for Primary Mammalian Endothelial Cells
Nucleofector® Basic Kit for Primary Mammalian Endothelial Cells

Basic Nucleofector® kit for primary mammalian endothelial cells, 25

CAS Number:  
Price
$1,333.13
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

In addition to various cell type-specific kits for primary endothelial cells, we also provide a cell group-specific basic Nucleofector® kit.

  • Optimization can be performed within 1 experiment
  • Detailed protocol guiding through the optimization procedure
  • Optional result fine tuning with help of our Scientific Support Team
  • Transfection efficiencies up to 90%
  • Viabilities up to 85%

This kit is suited to determine the optimal transfection conditions for any mammalian endothelial cell type that lacks a cell type specific optimized protocol for the Nucleofector® 2b device (or older generations I or II). In a first experiment a selection of different Nucleofector® Programs is tested in combination with the basic Nucleofector® solution for endothelial cells.

Applications: Kit suited for endothelial cells from different mammalian species and various organs
Already tested for human pulmonary artery endothelial cells.

Delivery information: Content: 2.25 ml Nucleofector® solution for primary mammalian endothelial cells, 0.5 ml Supplement 1, 30 µg pmaxGFP® Vector (0.5 μg/μl in 10 mM Tris pH 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.

Specifications

Specifications

  • Description:
    Basic Nucleofector® kit for primary mammalian endothelial cells
  • No. of reactions:
    25

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.