Order Entry
Canada
Orders LinkContactUsLinkComponent
Nucleofector® Human Stem Cell Kit 2

Nucleofector® Human Stem Cell Kit 2

Human stem cell Nucleofector® kit 2, 25
Catalog # CA10064-634
Supplier #:  VPH-5022
Nucleofector® Human Stem Cell Kit 2
Catalog # CA10064-634
CAS Number:  
Nucleofector® Human Stem Cell Kit 2
Nucleofector® Human Stem Cell Kit 2

Human stem cell Nucleofector® kit 2, 25

CAS Number:  
Price
$1,333.13
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. The Human Stem Cell Nucleofector® kit 2 is one of our two kits suited for efficient transfection of primary human stem cells.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection

Due to significant diversity between different human stem cell clones, we have developed two Nucleofector® kits (1 and 2) and an easy-to-use Nucleofector® starter kit. The Human Stem Cell Nucleofector® starter kit is used to determine which one of our two Nucleofector® kits is best suited for transfecting a specific primary human stem cell clone in Nucleofector® 2b device. It is suitable for 18 optimization reactions.

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.

Delivery information: Content: 0.9 ml Human stem cell Nucleofector® solution 1, 0.9 ml human stem cell Nucleofector® solution 2, 2×0.2 ml supplement 1, 2×30 µg pmaxGFP® vector (0.5 μg/μl in 10 mM Tris pH 8.0), 20 aluminum cuvette (100 µl), 20 single use pipettes.

Documents

Specifications

Specifications

  • Description:
    Human stem cell Nucleofector® kit 2
  • No. of reactions:
    25

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.