Order Entry
Canada
ContactUsLinkComponent
Nucleofector® Chicken Neuron Kit
Nucleofector® Chicken Neuron Kit
Catalog # CA10064-448
CAS Number:  
undefined
Nucleofector® Chicken Neuron Kit
Catalog # CA10064-448
Supplier Number:  VPG-1002
CAS Number:  

Specifications

  • Description:
    Chicken neuron Nucleofector® kit
  • Cat. no.:
    CA10064-448
  • No. of Reactions:
    25

Specifications

About this item

The Nucleofector® 2b system (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary chicken neurons.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection

The kit is available in different variants for either 10× or 25× or 4×25 reactions.

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.

Delivery: Content: 2.25 ml Nucleofector® solution for chicken neurons, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.