Nucleofector® Mouse Neuron Kit
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary mouse neurons.
- High transfection efficiencies for primary cells
- Cells maintain functionality post transfection
Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
Delivery information: Content: 2.25 ml Nucleofector® solution for mouse neuron, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.
Documents
Amaxa® Mouse Neuron Nucleofector® Kit
Additional Product Details
Specifications
Specifications
- Description:Mouse neuron Nucleofector® kit
- No. of reactions:25
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



