Order Entry
Canada
ContactUsLinkComponent
Nucleofector® Mouse Embryonic Fibroblast Kit 1
Nucleofector® Mouse Embryonic Fibroblast Kit 1
Catalog # CA10064-434
CAS Number:  
undefined
Nucleofector® Mouse Embryonic Fibroblast Kit 1
Catalog # CA10064-434
Supplier Number:  VPD-1004
CAS Number:  

Specifications

  • Description:
    Mouse embryonic fibroblast Nucleofector® kit 1
  • Cat. no.:
    CA10064-434
  • No. of Reactions:
    25

Specifications

About this item

The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection

The mouse embryonic fibroblast Nucleofector® kits are suitable for efficient transfection of primary mouse embryonic fibroblasts (MEF). Due to significant diversity between different MEF clones, we have developed two Nucleofector® Kits (1 and 2) and an easy-to-use Nucleofector® Starter Kit to determine which of the two kits is optimal for a specific MEF clone.

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.

Delivery: Content: 2.25 ml Nucleofector® solution for mouse embryonic fibroblast cells, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris pH 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.