Order Entry
Canada
ContactUsLinkComponent
Nucleofector® Human Dermal Fibroblast Kit
Nucleofector® Human Dermal Fibroblast Kit
  CA10064-428
 :  
undefined
Nucleofector® Human Dermal Fibroblast Kit
  CA10064-428
 :  VPD-1001
 :  

 

  • Description:
    Human dermal fibroblast Nucleofector® kit
  • Cat. no.:
    CA10064-428
  • No. of Reactions:
    25

 

 

The Nucleofector® 2b system (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary human fibroblasts, e.g. NHDF-neo.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.

 : Content: 2.25 ml Nucleofector® solution for human demal fibroblast cells, 0.5 ml supplement 4, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.