Nucleofector® Mouse Dendritic Cell Kit
CA10064-416
:
- Description:Mouse dendritic cell Nucleofector® kit
- Cat. no.:CA10064-416
- No. of Reactions:25
The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary mouse dendritic cells (immature or mature).
- High transfection efficiencies for primary cells
- Cells maintain functionality post transfection
Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
: Content: 2.25 ml Nucleofector® solution for mouse dendritic cells, 0.5 ml supplement 2, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.