ContactUsLinkComponent
Nucleofector® Mouse B Cell Kit
  CA10064-126
 :  
undefined
Nucleofector® Mouse B Cell Kit
  CA10064-126
 :  VPA-1010
 :  
  • Description:
    Mouse B cell Nucleofector® kit
  • Cat. no.:
    CA10064-126
  • No. of Reactions:
    25
  • Supplier No.:
    VPA-1010
The Nucleofector® 2b device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary mouse B cells.
  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
"Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
 : Content: 2.25 ml Nucleofector® solution for mouse B cells, 0.5 ml supplement 1, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.