Nucleofector® Mouse Macrophage Kit
Catalog # CA10064-124
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Mouse macrophage Nucleofector® kit
- Cat. no.:CA10064-124
- No. of Reactions:25
- Supplier No.:VPA-1009
Specifications
About this item
The Nucleofector® 2b system (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary mouse macrophages.
- High transfection efficiencies for primary cells
- Cells maintain functionality post transfection
The respective cell type-specific optimized protocol or recommendation is available under ‘Instructions’ or in our knowledge database. In this optimized protocol the best Nucleofection conditions are indicated. In addition, we share our experience and knowledge for treatment of this cell type.
Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.
Delivery: Content: 2.25 ml Nucleofector® solution for mouse macrophage cells, 0.5 ml supplement 2, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.