Order Entry
Canada
ContactUsLinkComponent
 
Nucleofector® Human Monocyte Kit
Catalog # CA10064-620
CAS Number:  
undefined
Nucleofector® Human Monocyte Kit
Catalog # CA10064-620
Supplier Number:  VPA-1007
CAS Number:  

Specifications

  • Description:
    Human monocyte Nucleofector® kit
  • Cat. no.:
    CA10064-620
  • No. of Reactions:
    25
  • Supplier No.:
    VPA-1007

Specifications

About this item

The Nucleofector® 2b system (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. This Nucleofector® kit is the optimal kit for efficient transfection of primary human monocytes.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low-throughput transfection in single cuvette format.

Delivery: Content: 2.25 ml Nucleofector® solution for human monocyte cells, 0.5 ml supplement 1, 30 µg pmaxGFP® vector (0.5 μg/μl in 10 mm tris ph 8.0), 25 aluminum cuvette (100 µl), 25 single use pipettes.