Nucleofector® Cell Line Optimization Kit for Nucleofector® Device
Catalog # CA10064-602
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Cell line optimization Nucleofector® kit for Nucleofector® device
- Cat. no.:CA10064-602
- No. of Reactions:18
- Supplier No.:VCO-1001N
Specifications
About this item
The cell line Optimization Nucleofector® kit is the ideal tool to determine the optimal transfection conditions for any difficult-to-transfect cell line that lacks an Optimized protocol for the Nucleofector® 2b device (or older generations I or II).
- Efficient transfection of virtually any difficult-to-transfect cell line
- Simple and rapid optimization completed within just one experiment
It enables you to conveniently determine the optimal Nucleofection condition of your cell line of interest within one experiment. The kit contains two different Cell Line Nucleofector® Solutions, V and L, each of which should be tested in combination with seven different Nucleofector® Programs. It is suitable for 18 optimization reactions.
Applications: Optimization of Nucleofection conditions of virtually any difficult-to-transfect cell line with the Nucleofector® II/2b device.
Delivery: Content:
0.9 ml cell line Nucleofector® solution L
0.9 ml cell line Nucleofector® solution V
2×0.2 ml supplement 1
2×30 µg pmaxGFP® Vector (0.5 μg/μl in 10 mM Tris pH 8.0)
20 Aluminum cuvette (100 µl)
20 Single use pipettes
0.9 ml cell line Nucleofector® solution L
0.9 ml cell line Nucleofector® solution V
2×0.2 ml supplement 1
2×30 µg pmaxGFP® Vector (0.5 μg/μl in 10 mM Tris pH 8.0)
20 Aluminum cuvette (100 µl)
20 Single use pipettes