Nucleofector® Cell Line Kit L
Cell line Nucleofector® kit L, 25
Supplier: LONZA PHARMA - BIOSCIENCE CA
Supplier #: VCA-1005
Nucleofector® Cell Line Kit L
Catalog # CA10064-600
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The cell line Nucleofector® kit L is one of our five Nucleofector® kits suited for transfecting cell lines in the Nucleofector® 2b device.
- High transfection efficiencies in difficult-to-transfect cell lines
This kit has been determined to be optimal for transfecting following cell types:
3T3-L1 ad
A-10
BHK-21
BxPC-3
DU145
EL4
FDC-P1
GH3 H9c2 (2-1)
HeLa S3
IMR-32
KG-1a
L-428 (DSMZ)
MDCK
MDCK II (ECACC)
MOLT-4
MV-4-11
NTERA-2 cl.D1
WEHI-231
Applications: Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells, low-throughput transfection in single cuvette format.
Delivery information: Content:
2.25 ml Nucleofector® solution R
0.5 ml supplement 1
30 µg pmaxGFP® Vector (0.5 μg/μl in 10 mM Tris pH 8.0)
25 Aluminum cuvette (100 µl)
25 Single use pipettes
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Cell line Nucleofector® kit L
- No. of reactions:25



