Nucleofector® Cell Line Kit C
Catalog # CA10064-598
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Cell line Nucleofector® kit C
- Size:25
- Cat. no.:CA10064-598
- Supplier No.:VCA-1004
Specifications
About this item
The cell line Nucleofector® kit C is one of our five Nucleofector® kits suited for transfecting cell lines in the Nucleofector® 2b device.
- High transfection efficiencies in difficult-to-transfect cell lines
This kit has been determined to be optimal for transfecting following cell types:
A2058
CCRF-CEM
MDA-MB-453
MEG-01
MG-63
NCI-H1299 [H1299]
NS0 (ECACC)
SK-BR-3
T2
U266B1
U-937
Applications: Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells, low-throughput transfection in single cuvette format.
Delivery: Content:
2.25 ml Nucleofector® solution R
0.5 ml supplement 1
30 µg pmaxGFP® Vector (0.5 μg/μl in 10 mM Tris pH 8.0)
25 Aluminum cuvette (100 µl)
25 Single use pipettes
2.25 ml Nucleofector® solution R
0.5 ml supplement 1
30 µg pmaxGFP® Vector (0.5 μg/μl in 10 mM Tris pH 8.0)
25 Aluminum cuvette (100 µl)
25 Single use pipettes