Order Entry
Canada
ContactUsLinkComponent
HMVEC-L – Human Lung Microvascular Endothelial Cells
HMVEC-L – Human Lung Microvascular Endothelial Cells
Catalog # CA10064-208
CAS Number:  
undefined
HMVEC-L – Human Lung Microvascular Endothelial Cells
Catalog # CA10064-208
Supplier Number:  CC-2527
CAS Number:  

Specifications

  • Description:
    HMVEC-L – Human lung microvascular endothelial cells
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-208
  • Supplier No.:
    CC-2527

Specifications

About this item

These lung microvascular endothelial cells include cells isolated from small vessels within normal lung tissue that are cultured to be ≥90% pure and specifically enriched for lung LECs and BECs.

  • Test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots

Lung microvascular cells, CC-2527 (HMVEC-L), are a mixed population of both blood (HMVEC-LBl) and lymphatic (HMVEC-LLy) endothelial cells.

Cryopreserved Clonetics® Lung Microvascular Endothelial Cells are guaranteed through 15 population doublings, express CD31/105, von Williebrand Factor VIII, are positive for acetylated low density lipoprotein uptake, and are PECAM positive. All cells test negative for mycoplasma, bacteria, yeast, and fungi.

Applications: Oncology, tumorigenesis, drug development, circulatory physiology, wound healing, lymph angiogenesis, immunology, basic research.

Delivery: A Certificate of Analysis is provided for each cell lot purchased.