Order Entry
Canada
ContactUsLinkComponent
NHDF-Neo – Human Dermal Fibroblasts, Neonatal
NHDF-Neo – Human Dermal Fibroblasts, Neonatal
Catalog # CA10064-198
CAS Number:  
undefined
NHDF-Neo – Human Dermal Fibroblasts, Neonatal
Catalog # CA10064-198
Supplier Number:  CC-2509
CAS Number:  

Specifications

  • Description:
    NHDF-Neo – Human dermal fibroblasts, neonatal
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-198
  • Supplier No.:
    CC-2509

Specifications

About this item

Human Dermal Fibroblasts are available from either adult skin (NHDF-Ad) or neonatal foreskin (NHDF-Neo).

They are derived from the dermis of normal human neonatal foreskin or adult skin. They are cryopreserved at the end of primary culture.

Lonza's human dermal fibroblasts are guaranteed through 15 population doublings when using FGM®-2 Growth Media. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. Dermal fibroblasts are characterized by morphological observation throughout serial passage.All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.

Applications: ECM protein analysis, wound healing, collagen metabolism, cosmetics, skin therapy/models.

Delivery: A Certificate of Analysis is provided for each cell lot purchased. Content: 1×cryopreserved ampule of NHDF-Neo containing ≥500000 cells.