Order Entry
Canada
ContactUsLinkComponent
HUVEC – Human Umbilical Vein Endothelial Cells, Single Donor, in EGM®-2, Lonza
HUVEC – Human Umbilical Vein Endothelial Cells, Single Donor, in EGM®-2, Lonza
Catalog # CA10065-208
CAS Number:  
undefined
HUVEC – Human Umbilical Vein Endothelial Cells, Single Donor, in EGM®-2, Lonza
Catalog # CA10065-208
Supplier Number:  C2517A
CAS Number:  

Specifications

  • Description:
    HUVEC – Human Umbilical Vein Endothelial cells, Single Donor, in EGM®-2
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10065-208
  • Supplier No.:
    C2517A

Specifications

About this item

Lonza's cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL® guaranteed through 5 PD).

  • Guaranteed through 15 population doublings
  • Test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots

HUVEC-XL size consists of 10 million cells/amp achieved by pooling donors. This option is designed to support studies that require large volume of cells while still offering cost-benefits to researchers.

Suitable HUVECs for research:
C2517A (single donor), C2519A (pooled donor), 00191027 (XL size) - Isolated in EGM®-2 media, in presence of growth factors such as VEGF, rhFGF, rhEGF, and others. Does not contain BBE.
C2517AS (single donor), C2519AS (pooled donor) - Pre-screened HUVECs in EGM®-2 media. Screened for angiogenesis markers and test positive for expression of eNOS, Axl, Tie-2 and VEGFr2.
CC-2517 (single donor), CC-2519 (pooled donor) - Both isolated in EGM® media, in absence of defined growth factors. Contains BBE. No exogenous VEGF.
CC-2935 (single donor) - Isolated in EGM® - Plus Media: Improved version of part number CC-2517. Offers faster proliferation rates than HUVEC isolated in EGM® Media. No exogenous VEGF.

Applications: Wound healing, angiogenesis, tissue engineering, inflammation, oncology, pharmacology, vascular modeling, transfection.

Delivery information: Content: 1×Cryopreserved ampule of HUVEC containing ≥500000 cells. A Certificate of Analysis is provided for each cell lot purchased.