ContactUsLinkComponent
Human Peripheral Blood CD4+ T Cells, Cryopreserved, 10 Million
Human Peripheral Blood CD4+ T Cells, Cryopreserved, 10 Million
Catalog # CA10065-194
CAS Number:  
undefined
Human Peripheral Blood CD4+ T Cells, Cryopreserved, 10 Million
Catalog # CA10065-194
Supplier Number:  2W-200
CAS Number:  
Specifications
  • Description:
    Human peripheral blood CD4+ T cells, cryopreserved
  • Size:
    10 million cells
  • Cat. no.:
    CA10065-194
  • Supplier No.:
    2W-200
Specifications
About this item
CD4+ T cells are also known as helper T cells because they act on other cells of the immune system to promote various aspects of the immune response,including immunoglobulin isotype switching and affinity maturation of the antibody response,macrophage activation and enhanced activity of natural killer cells and cytotoxic T cells.
  • Test negative for mycoplasma, bacteria, yeast, and fungi
  • Cells from a variety of donors available
Lonza CD4+T Cells are isolated from peripheral blood of healthy donors using negative immunomagnetic separation. Cryopreserved CD4+ T cells are guaranteed ≥ 90% pure for CD4+ T cells, as assessed by flow cytometry. They are guaranteed to contain ≥10 million viable cells per vial. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased. Cells are available from multiple donors.
Hematopoietic and immune research requires cells from different donors and tissue sources. That's why Lonza offers many different peripheral blood cell types. If you need different cell types from the same donor, you can either isolate from Lonza cryopreserved PBMCs or contact Customer Service to inquire about cells matched from the same donor. In addition, if you are interested in more naive T cells, unprocessed bone marrow, autologous bone marrow and whole blood from the same donor, bone marrow CD34+ cells, or cord blood CD34+ cells might be good solutions.
Lymphocyte Growth Medium is recommended for optimal performance. Please contact Scientific Support for thawing and culturing guidance. Lonza guarantees the performance of Clonetics®/Poietics® cells only if appropriate Clonetics®/Poietics® media and reagents are used exclusively and the recommended storage and use protocols are followed. Any modifications made to the recommended cell systems including the use of alternative media, reagents, or protocols, will void cell and media performance guarantees.
Delivery: Content: 1×Cryopreserved ampule of Human CD4+ T Cells containing ≥10 million cells.