Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Lucetta® 2 Luminometer

Lucetta® 2 luminometer, 100 - 240 V/50 - 60 Hz, 150×280×170 mm, ~2 kg
  76321-328
 :   Lonza
 :  AAL-1002
Lucetta® 2 Luminometer
  76321-328
 :  Lonza
Lucetta® 2 Luminometer
Lucetta® 2 Luminometer

Lucetta® 2 luminometer, 100 - 240 V/50 - 60 Hz, 150×280×170 mm, ~2 kg

 
$13,279.93
   
Each
 
 

The Lucetta® 2 luminometer is a single tube luminometer developed for the detection of bioluminescence and chemiluminescence. It can be used as a portable battery-operated instrument as well as with an external power supply and is suitable for detecting all luminescence.

  • Small, portable tube luminometer
  • Stand-alone instrument without PC requirements
  • High sensitivity and linearity
  • Specific measurement mode for running MycoAlert® Mycoplasma Detection Assays
  • Quick single read mode for unprocessed luminescence readings
  • High data storage capacity with maximum storage of up to 2000 single sample readings

Designed with a specific mode for using Lonza’s MycoAlert® Mycoplasma detection assays, it immediately provides assay results as CLEAN, BORDERLINE or CONTAMINATED sample.

The Lucetta® 2 Luminometer can also be used for many other luminescence-based assays, such as cell proliferation and cytotoxicity assays or luciferase reporter gene assays. For these assays, the instrument's single read mode applies.

Technical specifications:
Dimensions:150×280×170 mm (5.9×11.02×6.69")
Weight:Approx. 2 kg (4.4 lbs.)
Power supply input:100 to 240 V AC, 50 to 60 Hz
Output: 5.9 to 6 V DC/2.1 A
Power consumption (instrument):Max. 15 W
Operating temperature and humidity: 15 to 30 °C 80% humidity at 31 °C degreasing linearly to 50% at 40 °C, no condensation

Applications:
MycoAlert® Mycoplasma Detection Assays.
ATP assays, e.g. ViaLight® Cell Proliferation and Cytotoxicity Assay.
Adenylate Kinase assays, e.g. ToxiLight® Non-destructive Cytotoxicity Assay.
Luciferase reporter gene analysis.

 : Content: 1 Lucetta® 2 luminometer instrument.

 
  • Description:
    Lucetta® 2 luminometer
  • Dimensions:
    150×280×170 mm
  • Electrical:
    100 - 240 V/50 - 60 Hz
  • Weight:
    ~2 kg