Orders LinkContactUsLinkComponent
DNA QuantLadder
DNA QuantLadder
DNA QuantLadder, 125 µl
Catalog # CA12001-736
Supplier:   Lonza
Supplier #:  50334
DNA QuantLadder
Catalog # CA12001-736
Supplier:  Lonza
DNA QuantLadder
DNA QuantLadder
DNA QuantLadder, 125 µl
Price
$309.97
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Standard DNA ladders are ready-to-dilute prior to loading gel.
  • Plasmid free, no dilution needed
Plasmid-free to ensure minimal background. For all Lonza’s ladders, each band has an ideal 50% G-C content for rapid, precise sizing and sharp resolution. Our ladders are free from contaminating plasmid DNA, so they have minimal background, giving you the perfect gel the first time, every time.
Applications: DNA electrophoresis.
Delivery information: Content: DNA Quant ladder 125 µl.
Documents
Certificates
Search by Lot Number
Specifications
  • Description:
    DNA QuantLadder
  • Size:
    125 µl